Learn More
Coast WPH30R Waterproof and Dust Proof Headlamp
Catalog No. 17010211
Encompass
Change view
Click to view available options
Light Source:
LED
Catalog No. Light Source
17010211 LED
1 options
Catalog No. 17010211 Supplier Coast Supplier No. 30741
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
As the mid-size do-it-all model in our waterproof headlamp collection, the WPH30R is equally high-performance and multipurpose. Sealed tight to withstand full submersion, this IP68-built headlamp shines in five output modes spanning two beam colors: white and red.
  • Light body rolls in its bracket for directional control but can also detach to become a handheld light
  • Illumination that peaks at 1000 lumens with a max beam distance of 152 meters (498 feet)
  • Equipped with the COAST Ultra-Strap—a silicone-lined, non-slip breathable mesh band with an integrated buckle—the WPH30R can be worn, hung, or strapped to structures for lighting convenience
  • Featuring the COAST Dual Power system, this all-conditions headlamp runs for up to 23 hours on a single charge of its COAST ZITHION-X™ rechargeable lithium-ion battery or can operate on a CR123 disposable lithium battery
Specifications
Light Source LED
Voltage 3.7 V
Product Type General Purpose Lamp