missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wortmannin 98.0+%, TCI America™
Supplier: TCI America W000720MG
Specifications
| Wortmannin | |
| 237°C | |
| C23H24O8 | |
| 20 mg | |
| wortmannin, wartmannin, antibiotic sl-2052, unii-xva4o219qw, xva4o219qw, pi 3-kinase inhibitor, 1alpha,11alpha-11-acetyloxy-1-methoxymethyl-2-oxaandrosta-5,8-dieno 6,5,4-bc furan-3,7,17-trione, 1r,3r,5s,9r,18s-18-methoxymethyl-1,5-dimethyl-6,11,16-trioxo-13,17-dioxapentacyclo 10.6.1.0^ 2,10 .0^ 5,9 .0^ 15,19 nonadeca-2 10 ,12 19 ,14-trien-3-yl acetate, 1s,6br,9as,11r,11br-9a,11b-dimethyl-1-methyloxy methyl-3,6,9-trioxo-1,6,6b,7,8,9,9a,10,11,11b-decahydro-3h-furo 4,3,2-de indeno 4,5-h isochromen-11-yl acetate, wortmanin | |
| COC[C@H]1OC(=O)C2=COC3=C2[C@@]1(C)C1=C([C@@H]2CCC(=O)[C@@]2(C)C[C@H]1OC(C)=O)C3=O | |
| 428.44 | |
| CHEBI:52289 | |
| ≥98.0% (HPLC) |
| 19545-26-7 | |
| Yellow | |
| MFCD00133927 | |
| 3462 | |
| QDLHCMPXEPAAMD-QAIWCSMKSA-N | |
| (1R,3R,5S,9R,18S)-18-(methoxymethyl)-1,5-dimethyl-6,11,16-trioxo-13,17-dioxapentacyclo[10.6.1.0²,¹â°.0âµ,â¹.0¹âµ,¹â¹]nonadeca-2(10),12(19),14-trien-3-yl acetate | |
| 312145 | |
| 428.44 | |
| Crystalline Powder |
Chemical Identifiers
| 19545-26-7 | |
| 428.44 | |
| QDLHCMPXEPAAMD-QAIWCSMKSA-N | |
| 312145 | |
| (1R,3R,5S,9R,18S)-18-(methoxymethyl)-1,5-dimethyl-6,11,16-trioxo-13,17-dioxapentacyclo[10.6.1.0²,¹â°.0âµ,â¹.0¹âµ,¹â¹]nonadeca-2(10),12(19),14-trien-3-yl acetate |
| C23H24O8 | |
| MFCD00133927 | |
| wortmannin, wartmannin, antibiotic sl-2052, unii-xva4o219qw, xva4o219qw, pi 3-kinase inhibitor, 1alpha,11alpha-11-acetyloxy-1-methoxymethyl-2-oxaandrosta-5,8-dieno 6,5,4-bc furan-3,7,17-trione, 1r,3r,5s,9r,18s-18-methoxymethyl-1,5-dimethyl-6,11,16-trioxo-13,17-dioxapentacyclo 10.6.1.0^ 2,10 .0^ 5,9 .0^ 15,19 nonadeca-2 10 ,12 19 ,14-trien-3-yl acetate, 1s,6br,9as,11r,11br-9a,11b-dimethyl-1-methyloxy methyl-3,6,9-trioxo-1,6,6b,7,8,9,9a,10,11,11b-decahydro-3h-furo 4,3,2-de indeno 4,5-h isochromen-11-yl acetate, wortmanin | |
| CHEBI:52289 | |
| COC[C@H]1OC(=O)C2=COC3=C2[C@@]1(C)C1=C([C@@H]2CCC(=O)[C@@]2(C)C[C@H]1OC(C)=O)C3=O |
Safety and Handling
RTECSNumber : CB9641000
TSCA : No
Recommended Storage : Freezer (-20 °C)