missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WORTMANNIN
Description
Wortmannin, a fungal metabolite, is a potent and selective inhibitor of phosphatidylinositol 3-kinase.
Format/Formulation: Lyophilized
Specifications
Specifications
| Content And Storage | Store in freezer (-5 to -30°C). |
| Gene ID (Entrez) | 5290, 5291, 5293, 5294 |
| Purification Method | TLC |
| Shipping Condition | Dry Ice |
| Specific Inhibit or Target | PI3-Kinase |
| Sterility | Non-sterile |
| Quantity | 1 mg |
| Product Type | Wortmannin |
| Inhibitor Properties | Chemical |
| Molecular Weight (g/mol) | 428.4 |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.