missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ Workstation Storage Bin
GREENER_CHOICE

Catalog No. 03448400
Change view
Click to view available options
Color:
Clear
Dimensions (L x W x H):
18 x 12.25 x 6.5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Dimensions (L x W x H)
03448400 Clear 18 x 12.25 x 6.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03448400 Supplier Heathrow Scientific™ Supplier No. HS234523

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Workstation Storage Bin is a lab consumables holder and dispenser that allows you to organize your most commonly used laboratory items in one central location.

  • Acrylic unit has room to store gloves, Kim wipes™, and Parafilm™
  • Lidded bins to store tubes, tips, or other small items
  • Provides 5 compartments

Specifications

Type Storage Bin
Material Acrylic
Color Clear
Shape Rectangular
Autoclavable Not Autoclavable
For Use With (Application) Storing and organizing lab items in one place
Holds Laboratory disposables
Dimensions (L x W x H) 18 x 12.25 x 6.5 in.
Weight (English) 6 lb.
Weight (Metric) 2.7 kg