Learn More
Brady™ Workstation Scan and Print Suite Email Activation Code
Description
Scan Mode - Scan existing barcodes to capture data and transfer onto a new label template.CVS Mode - Scan the name of a CSV file to print a specified batch of labels. Custom Script Mode - Perform advanced label operations such as parse scanned data or serialize labels. Scalable Workflows - Expand workflows to additional workstations using the Export and Import feature. Quickly replicate labels in a lab for sample testing, for aliquots at a bio bank, or in a variety of other environments.
Specifications
Specifications
| Product Type | Software |
| For Use With (Equipment) | Brady Label Printers, Brady Scanners |
| Width (English) | 0.002 in. |
| Width (Metric) | N/A |
| Includes | Email Activation Code for Software |
| Operating System | Windows™ 7 (32- or 64-bit);Windows™ 7 Enterprise;Windows™ 7 Professional;Windows™ 7 SPI;Windows™ 7 Ultimate;Windows™ 8 (32- or 64-bit);Windows™ 8 Professional (32 or 64 bit);Windows™ 8.1 (32- or 64-bit);Windows™ 8.1 Enterprise;Windows™ 8.1 Professional;Windows™ 10 |
| Quantity | 1 Ea. |