missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Workstation Print Partner Software
Description
- Print Partner has no design capabilities, so it is deployable on multiple locations where only printing is required
- Users can only complete designated fields so you are always sure that your labels meet the specifications of you or your customers
- Runs as an app within the Brady™ Workstation platform or as a stand-alone application
- The simplified user interface only gives you the required functionality while limiting training for users
- Windows™ 7 Professional; Windows™ 7 Ultimate; Windows™ 7 Enterprise; Windows™ 8.1 Enterprise; Windows™ 8.1 Professional
- Single user license
Specifications
Specifications
| Product Type | Software |
| For Use With (Equipment) | BBP11 Label Printer, BBP33 Label Printer, Brady 1244, Brady 1344, Brady IP Series Label Printer, BradyPrinter 300PR+, Brady 2461/3481/6441 Series, Brady MVP Plus Series, Brady X Plus II Series Label Machine, BMP41 Label Printer, BMP71 Label Printer, Laser Printers, Ink Jet Printers, BMP53 Label Printer, BMP51 Label Printer |
| Width (English) | 7.6 in. |
| Format | CD |
| Memory | 200 MB, 1.5 Gigahertz (GHZ) Processor or Faster with 2 GB RAM |
| For Use With (Application) | Label printing |