missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Workstation Print Partner Software

Catalog No. 19124562
Change view
Click to view available options
Format:
CD
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
19124562 CD
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19124562 Supplier Brady™ Supplier No. BWSPPCD
Only null left

Provides a print-only software companion to Brady LabelMark™ label creation software. Brady™ Workstation Print Partner Software has no design capabilities, and to you print needed labels with pre-determined label content.

  • Print Partner has no design capabilities, so it is deployable on multiple locations where only printing is required
  • Users can only complete designated fields so you are always sure that your labels meet the specifications of you or your customers
  • Runs as an app within the Brady™ Workstation platform or as a stand-alone application
  • The simplified user interface only gives you the required functionality while limiting training for users
  • Windows™ 7 Professional; Windows™ 7 Ultimate; Windows™ 7 Enterprise; Windows™ 8.1 Enterprise; Windows™ 8.1 Professional
  • Single user license

Specifications

Product Type Software
For Use With (Equipment) BBP11 Label Printer, BBP33 Label Printer, Brady 1244, Brady 1344, Brady IP Series Label Printer, BradyPrinter 300PR+, Brady 2461/3481/6441 Series, Brady MVP Plus Series, Brady X Plus II Series Label Machine, BMP41 Label Printer, BMP71 Label Printer, Laser Printers, Ink Jet Printers, BMP53 Label Printer, BMP51 Label Printer
Width (English) 7.6 in.
Format CD
Memory 200 MB, 1.5 Gigahertz (GHZ) Processor or Faster with 2 GB RAM
For Use With (Application) Label printing