Learn More
Wood's metal, rods, Thermo Scientific™
Supplier:  Thermo Scientific Chemicals 388555000
Catalog No. AC388555000
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Wood's metal
Pb·Sn
500 g
Glass bottle
76093-98-6
MFCD00198175
647.28
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Harmful if inhaled.
May damage the unborn child. Suspected of damaging fertility.
GHS P Statement
Obtain special instructions before use.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid contact during pregnancy/while nursing.
GHS Signal Word: Warning
SDS Documents