missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Wood Dynamics Carts

Catalog No. S96176
Change view
Click to view available options
Weight (English):
4.6 lb.
5.7 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (English)
S96176 5.7 lb.
S96175 4.6 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Catalog No. S96176 Supplier Eisco™ Supplier No. PH0350B

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Ideal for demonstrating Newton's laws of motion.

  • Carts made with low-friction ball-bearing wheels
  • Top surface can be loaded with additional masses
  • One cart in each set is equipped with an exploder piston, activated by a trigger on top that can be preset to three positions
  • Spring-steel hoop bumpers used for collision and energy experiments

Order Info

Sold as a set of two carts.

Specifications

Product Type Dynamics Trolley
Material Wood
For Use With (Application) Demonstrate Newton's laws of motion
Dimensions (L x W x H) 14.1 x 10.6 x 4.7 in. (36 x 27 x 12 cm)
Height (English) 4.7 in.
Height (Metric) 12 cm
Length (English) 14.1 in.
Length (Metric) 36 cm
Width (English) 10.6 in.
Width (Metric) 27 cm
Weight (English) 5.7 lb.
Weight (Metric) 2.6 kg
Show More Show Less