Learn More
Eisco™ Wood Dynamics Carts
Ideal for demonstrating Newton's laws of motion.
Supplier:  Eisco™ PH0350A
 View more versions of this product
Catalog No. S96175
Only null left
Alert:
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Description
Description
  • Carts made with low-friction ball-bearing wheels
  • Top surface can be loaded with additional masses
  • One cart in each set is equipped with an exploder piston, activated by a trigger on top that can be preset to three positions
  • Spring-steel hoop bumpers used for collision and energy experiments
Order Info
Sold as a set of two carts.
Specifications
Chemical Identifiers
Dynamics Trolley
Wood
Demonstrate Newton's laws of motion
Specifications
Dynamics Trolley
Demonstrate Newton's laws of motion
13.3 x 11.4 x 3.9 in. (34 x 29 x 10 cm)
10 cm
34 cm
29 cm
2.09 kg
Wood
Steel hoop bumpers
3.9 in.
13.3 in.
11.4 in.
4.6 lb.
Product Suggestions
Product Suggestions