missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Women's Navy Flame Resistant Flex Knit Long Sleeve T-Shirt

Catalog No. 17204357
Change view
Click to view available options
Size:
2X-Large (Long)
Large (Long)
X-Large (Long)
Small
Medium
Large
X-Large
2X-Large
3X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17204357 Small
17204048 Medium
17204047 Large
17204232 X-Large
17204460 2X-Large
17204227 3X-Large
17204142 Large (Long)
17204356 X-Large (Long)
17204077 2X-Large (Long)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 17204357 Supplier Bulwark™ Supplier No. STT3NV RG S

Women's Navy Flame Resistant Flex Knit Long Sleeve T-Shirt gives you the stretch of your favorite T-shirt while keeping you cool and dry.

  • Bulwark FR Women's Flex Knit Tee gives you the stretch of your favorite T-shirt while keeping you cool and dry.
  • The lightweight fabric flexes for added comfort and range of movement, curved flatlock seams for optimal fit, and a dropped back hem for a flattering silhouette.
  • CAT 2/2112 left sleeve label leaves no question you're protected on the job site.
  • Certifications Level 1: NFPA 2112, NFPA 70E
  • Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
  • Hazard: Flash Fire, Arc Flash
  • Number of Pockets: 1
  • Primary Closure: Pull On
  • Wash Care: Home Wash
  • Fabric: Body: 6.75 oz. (230 g/m&superscript_2;) Flame Resistant Jersey, Contrast Panels: 6.75 oz. (230 g/m&superscript_2;) Flame Resistant Jersey
  • Blend: Body: 93% Cotton / 7% Spandex Elastane,Contrast Panels: 93% Cotton / 7% Spandex Elastane
  • Pocket: Single chest pocket
  • Features: Contrasting pattern on sleeve for athletic look, Lightweight stretch fabric flexes with your body for added comfort and range of movement, Dropped back hem presents a flattering silhouette
  • Protection: ATPV Arc Rating: 8.3
  • Country of Origin: Imported, Made in USA Fabric
  • Gender: Female
  • Brand: Bulwark™

Specifications

Size Small
Gender Women's
Material Cotton, Spandex
Color Navy
Neck Style Hemmed Crew
Sleeve Style Long Sleeve
Construction Knit
Closure Type Pull On
Disposable Reusable
No. of Pockets 1
Hazard Risk Category Category 2
Arc Flash Rating 8.3
Certifications/Compliance NFPA 2112, NFPA 70E
Show More Show Less