missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ Women's Navy Flame Resistant Flex Knit Button Down Shirt
Women's Navy Flame Resistant Flex Knit Button Down Shirt combines the crisp, professional look of an office work shirt and the comfort of your favorite T-shirt. Lightweight stretch fabric and side seam gussets flex with your body for added comfort and range of movement.
Supplier: Bulwark™ STG5NVRGM
Description
- The best of two hard-working worlds, this lightweight FR Flex Knit combines the crisp, professional look of an office work shirt and the comfort of your favorite T-shirt.
- Lightweight stretch fabric and side seam gussets flex with your body for added comfort and range of movement.
- Two chest pockets with button flaps allows convenient storage.
- Easy-to-spot CAT 2/2112 label on the left sleeve leave no question that you're protected on the job site.
- Certifications Level 1: NFPA 2112, NFPA 70E, ASTM F1506
- Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
- Hazard: Arc Flash, Flash Fire
- Number of Pockets: 2
- Primary Closure: Button
- Wash Care: Industrial Laundry - Light Soil
- Fabric: Body: 6.75 oz. (230 g/m&superscript_2;) Flame Resistant Jersey
- Blend: Body: 93% Cotton / 7% Spandex Elastane
- Collar: Banded Collar
- Pocket: Two chest pockets with flaps and button closure
- Features: Banded collar, Full underarm gusset for added mobility, Lightweight, stretch fabric, Two chest pockets with flap and sewn in pencil stall
- Protection: ATPV Arc Rating: 8.3
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Feminine Fit
- Gender: Female
- Brand: Bulwark™
Specifications
| Medium | |
| Cotton, Spandex | |
| Collar | |
| Knit | |
| Reusable | |
| Category 2 | |
| NFPA 2112, NFPA 70E, ASTMF1506 |
| Women's | |
| Navy | |
| Long Sleeve | |
| Button | |
| 2 | |
| 8.3 |