missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Women's Navy Flame Resistant Flex Knit Button Down Shirt

Catalog No. 17204273
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17204273 Small
17204463 Medium
17204256 Large
17204192 X-Large
17204468 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17204273 Supplier Bulwark™ Supplier No. STG5NV RG S

Women's Navy Flame Resistant Flex Knit Button Down Shirt combines the crisp, professional look of an office work shirt and the comfort of your favorite T-shirt. Lightweight stretch fabric and side seam gussets flex with your body for added comfort and range of movement.

  • The best of two hard-working worlds, this lightweight FR Flex Knit combines the crisp, professional look of an office work shirt and the comfort of your favorite T-shirt.
  • Lightweight stretch fabric and side seam gussets flex with your body for added comfort and range of movement.
  • Two chest pockets with button flaps allows convenient storage.
  • Easy-to-spot CAT 2/2112 label on the left sleeve leave no question that you're protected on the job site.
  • Certifications Level 1: NFPA 2112, NFPA 70E, ASTM F1506
  • Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
  • Hazard: Arc Flash, Flash Fire
  • Number of Pockets: 2
  • Primary Closure: Button
  • Wash Care: Industrial Laundry - Light Soil
  • Fabric: Body: 6.75 oz. (230 g/m&superscript_2;) Flame Resistant Jersey
  • Blend: Body: 93% Cotton / 7% Spandex Elastane
  • Collar: Banded Collar
  • Pocket: Two chest pockets with flaps and button closure
  • Features: Banded collar, Full underarm gusset for added mobility, Lightweight, stretch fabric, Two chest pockets with flap and sewn in pencil stall
  • Protection: ATPV Arc Rating: 8.3
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Feminine Fit
  • Gender: Female
  • Brand: Bulwark™

Specifications

Size Small
Gender Women's
Material Cotton, Spandex
Color Navy
Neck Style Collar
Sleeve Style Long Sleeve
Construction Knit
Closure Type Button
Disposable Reusable
No. of Pockets 2
Hazard Risk Category Category 2
Arc Flash Rating 8.3
Certifications/Compliance NFPA 2112, NFPA 70E, ASTM F1506
Show More Show Less