missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ Women's Khaki Flame Resistant Flex Knit Long Sleeve T-Shirt
Women's Khaki Flame Resistant Flex Knit Long Sleeve T-Shirt gives you the stretch of your favorite T-shirt while keeping you cool and dry.
$124.80 - $124.80
Specifications
| Gender | Women's |
|---|---|
| Material | Cotton, Spandex |
| Color | Khaki |
| Neck Style | Hemmed Crew |
| Sleeve Style | Long Sleeve |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
17-204-334
|
Bulwark™
STT3KHRGS |
Small |
Each for $124.80
|
|
|||||
|
17-204-010
|
Bulwark™
STT3KHRGM |
Medium |
Each for $124.80
|
|
|||||
|
17-204-245
|
Bulwark™
STT3KHRGL |
Large |
Each for $124.80
|
|
|||||
|
17-204-343
|
Bulwark™
STT3KHRGXL |
X-Large |
Each for $124.80
|
|
|||||
|
17-204-419
|
Bulwark™
STT3KHRGXXL |
2X-Large |
Each for $124.80
|
|
|||||
|
17-204-002
|
Bulwark™
STT3KHRG3XL |
3X-Large |
Each for $124.80
|
|
|||||
|
17-204-086
|
Bulwark™
STT3KHLNL |
Large (Long) |
Each for $124.80
|
|
|||||
|
17-203-993
|
Bulwark™
STT3KHLNXL |
X-Large (Long) |
Each for $124.80
|
|
|||||
|
17-203-998
|
Bulwark™
STT3KHLNXXL |
2X-Large (Long) |
Each for $124.80
|
|
|||||
Description
- Bulwark FR Women's Flex Knit Tee gives you the stretch of your favorite T-shirt while keeping you cool and dry.
- The lightweight fabric flexes for added comfort and range of movement, curved flatlock seams for optimal fit, and a dropped back hem for a flattering silhouette.
- CAT 2/2112 left sleeve label leaves no question you're protected on the job site.
- Certifications Level 1: NFPA 2112, NFPA 70E
- Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
- Hazard: Flash Fire, Arc Flash
- Number of Pockets: 1
- Primary Closure: Pull On
- Wash Care: Home Wash
- Fabric: Body: 6.75 oz. (230 g/m&superscript_2;) Flame Resistant Jersey, Contrast Panels: 6.75 oz. (230 g/m&superscript_2;) Flame Resistant Jersey
- Blend: Body: 93% Cotton / 7% Spandex Elastane, Contrast Panels: 93% Cotton / 7% Spandex Elastane
- Pocket: Single chest pocket
- Features: Contrasting pattern on sleeve for athletic look, Lightweight stretch fabric flexes with your body for added comfort and range of movement, Dropped back hem presents a flattering silhouette
- Protection: ATPV Arc Rating: 8.3
- Country of Origin: Imported, Made in USA Fabric
- Gender: Female
- Brand: Bulwark™
Specifications
| Women's | |
| Khaki | |
| Long Sleeve | |
| Reusable | |
| Category 2 | |
| NFPA 2112, NFPA 70E |
| Cotton, Spandex | |
| Hemmed Crew | |
| Knit | |
| 1 | |
| 8.3 |