missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Women's Khaki Flame Resistant Flex Knit Button Down Shirt

Catalog No. 17204404
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17204404 Large
17204163 Small
17204278 Medium
17103747 X-Large
17103746 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17204404 Supplier Bulwark™ Supplier No. STG5KH RG L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Women's Khaki Flame Resistant Flex Knit Button Down Shirt combines the crisp, professional look of an office work shirt with the comfort of your favorite T-shirt.

  • Lightweight stretch fabric and side seam gussets flex with your body for added comfort and range of movement
  • Two chest pockets with button flaps allows convenient storage
  • Sewn in pencil stall and banded collar
  • Easy-to-spot CAT 2/2112 label on the left sleeve leave no question that you're protected on the job site
  • Hazard : Arc Flash, Flash Fire
  • Wash Care: Industrial Laundry - Light Soil
  • Fabric: Body: 6.75 oz. (230 g/m2) Flame Resistant Jersey
  • Blend: Body: 93% Cotton/7% Spandex Elastane
  • Protection: ATPV Arc Rating: 8.3
  • Country of Origin: Imported, Made in USA Fabric

Specifications

Size Large
Gender Women's
Material Cotton, Spandex
Color Khaki
Neck Style Collar
Sleeve Style Long Sleeve
Construction Knit
Closure Type Button
Disposable Reusable
No. of Pockets 2
Hazard Risk Category Category 2
Arc Flash Rating 8.3
Certifications/Compliance NFPA 2112, NFPA 70E, ASTM F1506
Show More Show Less