missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ Women's Khaki Flame Resistant Flex Knit Button Down Shirt
Women's Khaki Flame Resistant Flex Knit Button Down Shirt combines the crisp, professional look of an office work shirt with the comfort of your favorite T-shirt.
Supplier: Bulwark™ STG5KHRGM
Description
- Lightweight stretch fabric and side seam gussets flex with your body for added comfort and range of movement
- Two chest pockets with button flaps allows convenient storage
- Sewn in pencil stall and banded collar
- Easy-to-spot CAT 2/2112 label on the left sleeve leave no question that you're protected on the job site
- Hazard : Arc Flash, Flash Fire
- Wash Care: Industrial Laundry - Light Soil
- Fabric: Body: 6.75 oz. (230 g/m2) Flame Resistant Jersey
- Blend: Body: 93% Cotton/7% Spandex Elastane
- Protection: ATPV Arc Rating: 8.3
- Country of Origin: Imported, Made in USA Fabric
Specifications
| Medium | |
| Cotton, Spandex | |
| Collar | |
| Knit | |
| Reusable | |
| Category 2 | |
| NFPA 2112, NFPA 70E, ASTM F1506 |
| Women's | |
| Khaki | |
| Long Sleeve | |
| Button | |
| 2 | |
| 8.3 |