missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Women's Khaki Flame Resistant Flex Knit Button Down Shirt

Catalog No. 17204163
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17204163 Small
17204278 Medium
17204404 Large
17103747 X-Large
17103746 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17204163 Supplier Bulwark™ Supplier No. STG5KH RG S

Women's Khaki Flame Resistant Flex Knit Button Down Shirt combines the crisp, professional look of an office work shirt with the comfort of your favorite T-shirt.

  • Lightweight stretch fabric and side seam gussets flex with your body for added comfort and range of movement
  • Two chest pockets with button flaps allows convenient storage
  • Sewn in pencil stall and banded collar
  • Easy-to-spot CAT 2/2112 label on the left sleeve leave no question that you're protected on the job site
  • Hazard : Arc Flash, Flash Fire
  • Wash Care: Industrial Laundry - Light Soil
  • Fabric: Body: 6.75 oz. (230 g/m2) Flame Resistant Jersey
  • Blend: Body: 93% Cotton/7% Spandex Elastane
  • Protection: ATPV Arc Rating: 8.3
  • Country of Origin: Imported, Made in USA Fabric

Specifications

Size Small
Gender Women's
Material Cotton, Spandex
Color Khaki
Neck Style Collar
Sleeve Style Long Sleeve
Construction Knit
Closure Type Button
Disposable Reusable
No. of Pockets 2
Hazard Risk Category Category 2
Arc Flash Rating 8.3
Certifications/Compliance NFPA 70E, NFPA 2112, ASTM F1506
Show More Show Less