missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark Women's FR/CP Lab Coat
Flame resistant/chemical protective lab coat brings together the latest innovations in lab safety to protect wearers against multiple hazards.
Supplier: Bulwark KNR7RBS0R
Description
Flame resistant/chemical protective lab coat brings together the latest innovations in lab safety to protect wearers against multiple hazards.
- Lightweight and breathable fire resistant/chemical protective lab coat.
- Black lay-down collar for easy recognition as a CP lab coat.
- Splash resistant pass-through for inner pocket access.
- Flame-resistant 4.5 oz. Nomex IIIA fabric prevents clothing ignition during short-duration thermal incidents and emergency exposure to flame.
- Repels small amounts of liquid chemicals that are either dropped or splashed on the fabric, resisting penetration and wicking of an inadvertent, secondary liquid chemical splash.
- Concealed snap front neck closures.
- One double-stitched chest pocket and two double-stitched lower pockets with flaps
- Arc Rating ATPV 5.6 cal/cm2
- Category 1 Protection
- NFPA 2112 Compliant
Specifications
| Chemical Resistant Lab Coat | |
| Women's | |
| Royal Blue | |
| Knit | |
| Non-sterile | |
| Regular | |
| Lay-down |
| Small | |
| Aramid Fiber | |
| Snaps | |
| Reusable | |
| Flame, Chemicals | |
| 3 | |
| Category 1, NFPA 2112 |