missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Women's Curvy Fit Flame Resistant Jean with Stretch

Catalog No. p-200000053
Change view
Click to view available options
Size:
0
10
12
14
16
18
2
20
22
24
4
6
8
Size (English) Inseam:
30 in.
32 in.
35 in.
31 product options available for selection
Product selection table with 31 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Size (English) Inseam
17204326 0 35 in.
17204344 2 35 in.
17204237 4 30 in.
17204238 4 32 in.
17204426 4 35 in.
17204389 6 30 in.
17204247 6 32 in.
17204348 6 35 in.
17204122 8 30 in.
17204250 8 32 in.
17204351 8 35 in.
17203954 10 30 in.
17204126 10 32 in.
17204454 10 35 in.
17204294 12 30 in.
17204216 12 32 in.
17204251 12 35 in.
17203948 14 30 in.
17204353 14 32 in.
17204123 14 35 in.
17204346 16 30 in.
17203952 16 32 in.
17204041 16 35 in.
17204141 18 30 in.
17204365 18 32 in.
17204149 18 35 in.
17204457 20 30 in.
17204158 20 32 in.
17204362 20 35 in.
17204252 22 35 in.
17204042 24 35 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 31 options available.
31 options
Catalog No. 17204326 Supplier Bulwark™ Supplier No. PSJCSD 00 35U
Only null left

Women's Curvy Fit Flame Resistant Jean with Stretch feel like an everyday jean. Light hand sanding and whiskering gives a more fashionable appearance. Felled inseam and seat seam adds durability. Contoured waistband provides a more comfortable fit.

  • Lightweight fabric with stretch that makes this work jean feel like an everyday jean.
  • Light hand sanding and whiskering gives a more fashionable appearance.
  • Felled inseam and seat seam adds durability.
  • Contoured waistband provides a more comfortable fit.
  • Metal button closure (metal is not exposed to the skin).
  • Two patch on spade shaped hip pockets.
  • Scoop front pockets with watch pocket on right side.
  • Zipper fly closure.
  • Certifications Level 1: NFPA 70E, NFPA 2112
  • Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
  • Hazard: Arc Flash, Flash Fire
  • Number of Pockets: 5
  • Primary Closure: Concealed Button
  • Wash Care: Industrial Laundry - Light Soil
  • Fabric: Body: 12.5 oz. (425 g/m&superscript_2;) Flame Resistant Denim
  • Blend: Body: 98% Cotton / 2% Spandex Elastane
  • Waistband: Contoured waistband with stretch interlining
  • Features: Contoured waistband, Stretch fabric, Hand sanded fabric with whiskering, Felled inseam and seat seam
  • Protection: ATPV Arc Rating: 21
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Curvy
  • Gender: Female
  • Brand: Bulwark™

Specifications

Size 0
Gender Women's
Material Cotton, Polyester, Spandex, Polyester, Spandex
Color Denim
Seam Type Stitched
No. of Pockets 4
Closure Type Concealed Button
Disposable Reusable
Hazard Risk Category Category 2
Arc Flash Rating 21
Certifications/Compliance NFPA 70E, NFPA 2112
Size (English) Inseam 35 in.
Show More Show Less