missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Women's Charcoal Plaid Flame Resistant Flex Knit Button Down Shirt

Catalog No. 17204410
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17204410 Small
17204267 Medium
17204171 Large
17203986 X-Large
17204416 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17204410 Supplier Bulwark™ Supplier No. STG3CH RG S

Women's Charcoal Plaid Flame Resistant Flex Knit Button Down Shirt combines the crisp, professional look of an office work shirt and the comfort of your favorite T-shirt.

  • The best of two hard-working worlds, this lightweight FR Flex Knit combines the crisp, professional look of an office work shirt and the comfort of your favorite T-shirt.
  • Lightweight stretch fabric flexes with your body for added comfort and range of movement.
  • Button-down collar keeps you looking crisp and sharp all day.
  • Easy-to-spot CAT 2/2112 label on the left sleeve leave no question that you're protected on the job site.
  • Certifications Level 1: ASTM F1506, NFPA 2112, NFPA 70E
  • Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
  • Hazard: Arc Flash, Flash Fire
  • Number of Pockets: 1
  • Primary Closure: Button
  • Wash Care: Industrial Laundry - Light Soil
  • Fabric: Body: 6.75 oz. (230 g/m&superscript_2;) Flame Resistant Jersey
  • Blend: Body: 93% Cotton / 7% Spandex Elastane
  • Features: Button down collar, Single chest pocket with pencil stall, Lightweight, stretch fabric
  • Protection: ATPV Arc Rating: 8.3
  • Country of Origin: Imported, Made in USA Fabric
  • Gender: Female
  • Brand: Bulwark™

Specifications

Size Small
Gender Women's
Material Cotton, Nylon
Color Charcoal
Neck Style Collar
Sleeve Style Long Sleeve
Construction Knit
Closure Type Button
Disposable Reusable
No. of Pockets 1
Hazard Risk Category Category 2
Arc Flash Rating 8.3
Certifications/Compliance ASTM F1506, NFPA 2112, NFPA 70E
Show More Show Less