missing translation for 'onlineSavingsMsg'
Learn More

Salicylic Acid, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical

Catalog No. 18606150
Change view
Click to view available options
Quantity:
125 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606150 45 kg Fiber Drum
18606143 125 g Amber Glass Bottle
18606144 6 x 125 g Amber Glass Bottle
18606147 2.5 kg Poly Pail
18606146 6 x 500 g Amber Glass Bottle
18606148 4 x 2.5 kg Poly Pail
18606149 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18606150 Supplier Spectrum Chemical Mfg Cor Supplier No. S100545KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S1005, 69-72-7, C7H6O3

Spectrum™ Chemical Salicylic Acid, Crystal, Reagent, ACS is an alpha-hydroxybenzoic acid and a colorless crystalline acid that is organic and commonly used in organic synthesis. It can be found occurring naturally in unripe fruits and vegetables and most commonly in the white willow tree. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 69-72-7
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00002439
InChI Key YGSDEFSMJLZEOE-UHFFFAOYSA-N
IUPAC Name 2-hydroxybenzoic acid
SMILES OC(=O)C1=CC=CC=C1O

Specifications

Melting Point 158°C to 161°C
CAS Max % 1
Quantity 45 kg
Packaging Fiber Drum
Residue after Ignition 0.0001
Percent Purity 99%
Grade ACS