missing translation for 'onlineSavingsMsg'
Learn More

Air-Tite Wing Web Vaccinators for Fowl Pox

Catalog No. 361053886
Change view
Click to view available options
Animal:
Small Bird
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Animal
361053886 Small Bird
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361053886 Supplier Air-Tite Supplier No. VBEIGE2

missing translation for 'validMainframeMsg'

Air-Tite Wing Web Vaccinators for Fowl Pox are high quality, economical, and reliable adult bird vaccinators

  • The Pox vaccinator is not a precision delivery device
  • Contains two 1.22 mm grooves for adult bird dosing of 0.01 mL

Specifications

Animal Small Bird
Type Vaccinators
Color Beige
Capacity (Metric) Dosing of 0.01 mL