missing translation for 'onlineSavingsMsg'
Learn More

Mopec Windowed White Evidence Envelopes

Catalog No. 22446316
Change view
Click to view available options
Dimensions (L x W):
6 x 9 in.
9 x 12 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22446316 9 x 12 in.
22446257 6 x 9 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22446316 Supplier Mopec Supplier No. FS422

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for secure evidence handling and quick visual identification, these white envelopes include a clear viewing window that allows contents or labels to be seen without opening the package. This helps preserve evidence integrity while making routine identification and handling more efficient.

Construction

  • Viewing window on one side
  • Printed write-on area with chain of custody

Quantity

  • 100 per package

Dimensions

  • FS421: 6 x 9 in.
  • FS422: 9 x 12 in.

Specifications

Dimensions (L x W) 9 x 12 in.
Product Type Evidence Envelopes
Quantity 100/Pk.