Learn More
Chemplex Industries WINDOW FRAME0.16 MIL 3.0IN DIA
Supplier: Chemplex Industries 3018
CHEMPLEX SPECTROMEMBRANE(TM) Thin-Film Sample Support Window Frames, Prolene(TM), 4.0 u (0.16 mil), 3.0 Dia., In.