Learn More
Contec Window Cleaning Squeegee and Replacement Parts
Catalog No. 17103024
Change view
Click to view available options
Product Type:
Replacement Blade
Single-blade Squeegee
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
17103024 Replacement Blade
17103023 Single-blade Squeegee
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17103024 Supplier Contec Supplier No. WCSQB018
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Stainless steel squeegee is lightweight, easy to use, and compatible with QuickConnect handles for hard-to-reach areas. Replacement rubber blade sold separately.
Features Benefits:
  • Lightweight, stainless-steel tool and rubber grip
  • Rubber grip prevents tool from slipping when used with disinfectants
  • Lightweight to clean tall windows and hard to reach areas
  • Maneuvers easily to use vertically and horizontally
Compatibility:
  • Compatible with all Contec's QuickConnect handles making windows easier to reach.
  • All components of the squeegee are compatible with common cleanroom cleaners and disinfectants
Press Down Handle Clip
  • Allows for easy installation preventing the rubber blade from moving during use
Screw-free channel
  • Blades to be replaced quickly and easily saving time
Autoclavable
  • Squeegee can be autoclaved at 121°C for 30 minutes
  • Ideal for use in all areas of the cleanroom
Specifications
Product Type Replacement Blade
Color-coded No
Sterility Non-sterile
Autoclavable Not Autoclavable
Color Black
Material Rubber