missing translation for 'onlineSavingsMsg'
Learn More

Contec Window Cleaning Solution

Catalog No. 17103021
Change view
Click to view available options
Color-coded:
No
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color-coded
17103021 No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17103021 Supplier Contec Supplier No. SBT32WC

The Window Cleaning Solution removes cleanroom residues from glass and hard surfaces. It contains 3% IPA, 0.2% ammonium hydroxide, and 2.9% surfactant in DI water.

  • Unique blend of solution solubilizes residues allowing them to be easily removed from surfaces
  • Low IPA level means little to no VOC's compared to 70% IPA.
  • Trigger and recloseable cap tailored to meet end-user preferences and SOP guidelines.
  • Irradiated solution filtered through 0.2 micron eliminates the need for in-house filtration
  • Prefiltered, packaged and irradiated for easy introduction into critical environments

Specifications

Product Type Solution
Color-coded No
Form Liquid
pH 11
Scent Original
Sterility Sterile