missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Wilmad™ Rotor and Caps for Bruker MAS Probe

Eliminate the need to solvate with MAS rotors manufactured from the highest purity Zirconia. Rotors provide the ultimate solution for analysis of solid samples.

Supplier:  Bel-Art™ 660000099

 View more versions of this product

Catalog No. 16800199

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

  • Caps fitted with O-rings for improved sealing
  • MAS Probe and caps are 100% compatible for most solid state NMR spectrometers
  • Thoroughly inspected before and after the precision machining process to ensure there are no material irregularities
  • Spin testing is performed to only the highest specified spinning speed, assuring performance without overspinning the rotor
Specifications

Specifications

Bruker MAS Probe (4.0 mm)
Five Rotors, Ten Kel-F Caps and Three Torlon Caps
WP-501-4180-SET-5
-100°C to +200°C
Cap
Various
Vmax = 18 kHz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions