Learn More
Bel-Art™ Wilmad™ Pyrex™ MAS Rotor Inserts
Designed for air-sensitive samples and semi-solid samples such as gels or highly viscous liquids. Inserts allow samples to be sealed into an NMR rotor by heat-seal or epoxy.
$398.44
Specifications
Type Rotor
For Use With (Application) Designed for air-sensitive samples and semi-solid samples such as gels or highly viscous liquids.
Includes Rotor Insert
Material Borosilicate Glass
View More Specs
Products 1
Catalog Number Mfr. No. Diameter (Metric) Diameter (Metric) Inner Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Diameter (Metric) Inner Price Quantity & Availability  
16800134
View Documents Promotion Details
Bel-Art™
660000034
2.99 mm 2.24 mm
Each for $398.44
Only null left
Description
Description
  • A glove box may be required for sealing of air-sensitive samples
Specifications
Specifications
Rotor
Designed for air-sensitive samples and semi-solid samples such as gels or highly viscous liquids.
Rotor Insert
Borosilicate Glass