missing translation for 'onlineSavingsMsg'
Learn More

SP Wilmad-LabGlass™ High-Throughput NMR Tubes

Catalog No. 03411816
Change view
Click to view available options
Quantity:
100 Pack
50 Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03411816 100 Pack
03411815 50 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03411816 Supplier SP Supplier No. 669995702

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Double the writing area allows for easy sample marking and identification.

  • Average camber of 60 microns ensures spectrum clarity for small molecule (MW <250) samples up to 600 MHz.
  • 2x larger marking area eliminates cramped and unreadable sample names
  • Silkscreened writing area keeps sample names from easily wiping off
  • Designed for routine use in most low to mid field NMR spectrometers
  • Camber and concentricity values listed are the Total Indicator Reading (TIR)
  • Made from ASTM E438 Type 1 Class B glass
  • 100% inspected for surface defects and physical dimensions

Note: For experiments that involve cooling, heating, biological sample, multi-dimension, multi-nuclei or DNP techniques, please refer to Wilmad Precision NMR Tubes.

Specifications

Material Glass (Type 1, Class B)
Length (Metric) 178 mm
Product Type High Throughput NMR Tube
Diameter (Metric) Outer 5±0.02 mm
Length (English) 7 in.
Thickness (Metric) Wall 0.43 mm
Quantity 100 Pack