missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Shigemi™ Susceptibility Matched NMR Tubes
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Shigemi™ Susceptibility Matched NMR Tubes feature a susceptibility matched bottom and plunger to effectively reduce the boundary gradients at the edges of the Rf coil reducing the required sample volume by two thirds.
Supplier: Bel-Art™ SP Scienceware™ 660000028
Description
- Select tube based on solvent used: CDCL3, CD3OD, D2O, DMSO-d6
- Available for Bruker or Universal
Specifications
| Tube Set | |
| Bruker Spectometers | |
| 5 mm | |
| BMS-005B |
| Glass (Type 1, Class A) | |
| 8 mm (Bottom) | |
| Deuterium Oxide Matched |