missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Williams and Briggs Salt Mixture

Catalog No. MP290283702
Change view
Click to view available options
Quantity:
2 kg
50 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP290283702 2 kg
MP290283750 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP290283702 Supplier MP Biomedicals™ Supplier No. 0290283702

Salt mixture for research animal diet.

  • Salt Mixture
  • For research animal diet
  • Powder

Specifications

Product Type Animal Diet
For Use With (Application) Research animal diet
Quantity 2 kg
Storage Requirements Room Temperature