missing translation for 'onlineSavingsMsg'
Learn More
Learn More
United Scientific™ UniStore™ Reagent Bottles, Wide Mouth, PP
Wide Mouth Polypropylene Reagent Bottles are rigid, translucent and have fine contact clarity.
Supplier: United Scientific™ 33308
Description
UniStore™ Wide Mouth, PP, reagent bottles are manufactured from USP Class VI resin that is FDA-21CFR compliant.
- Excellent chemical resistance to most acids, bases, and alcohols.
- Polypropylene caps are included.
- Manufactured from USP Class VI resin that is FDA-21CFR compliant for food contact and free of heavy metals.
- Leakproof closure with integrated seal ring.
- Precision molded for consistent wall thickness with smooth interior and exterior surfaces.
- Certificates of Analysis for leakproof test details are available upon request.
- The 4000 mL bottle is leakproof at ambient temperature and pressure.
Specifications
| 250 mL (8.5 oz.) | |
| 8.5 oz. | |
| 33 mm | |
| 12 Pk. | |
| UniStore | |
| No | |
| 131 mm | |
| 5.2 in. | |
| 43 mm | |
| No |
| 250 mL | |
| Translucent | |
| Polypropylene | |
| Non-sterile | |
| Yes | |
| Wide | |
| Reagent Bottle | |
| Polypropylene | |
| 33 mm | |
| Autoclavable |