missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ VITLAB™ Wide Mouth Bottles with Screw Caps

Transparent and resilient, with a wide mouth for easy filling.

$321.16 - $321.16

Specifications

Color Transparent
Material LDPE
Quantity 12/Pk.
Sterility Non-sterile
Disposable Single-use
View More Specs

Products 1
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
14-379-362
View Documents
BRANDTECH™ VITLAB™
93989
Pack of 12 Each for $321.16
Only null left
 
Description

Description

  • Bottles crafted from transparent low-density polyethylene
  • Screw cap closures made of polypropylene
  • Flexible and transparent, suitable for storing various substances including powders and viscous materials
Specifications

Specifications

Transparent
12/Pk.
Single-use
Not Autoclavable
LDPE
Non-sterile
Storage and Transport Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'