missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ Whizard™ Defender™ 10 Cut-Resistant Gloves

Catalog No. 19167806D
Change view
Click to view available options
Size:
X-Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19167806D Large
19167806A X-Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19167806D Supplier Wells Lamont™ Supplier No. 135481

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Whizard™ Defender™ 10 Cut-Resistant Gloves with high performance fibers and stainless steel combination offer ANSI Cut Level A6 cut resistance. 10-gauge gloves are ideal for handling sharp surfaces, food preparation, and meat and poultry processing.

Patented combination of high-performance fibers and stainless steel creates superior cut resistant properties, eliminating the high cost of lacerations.

  • Standard cuff
  • Ambidextrous
  • Prewashed and preshrunk
  • 10 gauge: ANSI A6
  • Made in the USA

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance FDA CFR 21, ASTM F2992-15, ASTM D3884-09
Coating Material Uncoated
Color White, Blue
Cut Resistance Rating ANSI A5
Hand Ambidextrous
Latex Free Yes
Material Fibers, Stainless Steel Fiber
Product Type Cut-resistant Gloves
Size Large
For Use With (Application) Medium duty applications
Glove Cuff Color Blue
Glove Cuff Style Plain
Hem Color White
Model 135481
Series Whizard™ Defender™ 10
Yarn Gauge 10
Show More Show Less