missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Whitney™ Medical Solutions Safe-Keeper™ Biohazard Container
Space-saving, economical, disposable point of use contaminated waste containers for disposal of capillary tubes, pipette tips, etc.
Supplier: Whitney™ Medical Solutions BH6012
Description
- For disposal of capillary tubes, pipette tips, stir rods and other point-of-use items
- Maximize workspace with hanging biohazard disposal units
- Made of sturdy plastic-lined paperboard
- Solid-bottom design improves rigidity and waste capacity
- Safe-Keeper can be autoclaved or incinerated
- Accommodates up to 1100cc of material and comes with a Hook-Up™ self-adhesive hanger
- Made in USA
Chemical Identifiers
| 5.1 cm | |
| Plastic | |
| 2 in. |
| Autoclavable | |
| Disposal of capillary tubes, pipette tips, stir rods and other point-of-use items | |
| Paperboard |
Specifications
| 2250 cc | |
| 1.9 cm | |
| 5.1 cm | |
| Plastic | |
| 2 in. | |
| 0.75 in. | |
| Self-adhesive Hook |
| 2 x 2 x 0.75 in. (5.1 x 5.1 x 1.9 cm) | |
| 5.1 cm | |
| Autoclavable | |
| Disposal of capillary tubes, pipette tips, stir rods and other point-of-use items | |
| 2 in. | |
| Paperboard |