missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heathrow Scientific™ Bench-Top Keeper™ Biohazard Container
Description
- Made of sturdy paperboard with an internal plastic lining and webbed corners to contain moderate amounts of fluids
- Self-locking tops keep contents safely enclosed
- Small Keepers fold out to 4L x 4W x 7H in. (10 x 10 x 18 cm) and accommodate up to 1600 cc of material
- Large Keepers fold out to 6L x 6W x 10H in. (15 x 15 x 25 cm) and accommodate up to 5000 cc of material
- Autoclavable
- Made in USA
Specifications
Specifications
| Capacity (Metric) | 1.4 L |
| Capacity (English) | 1400 cc |
| Color | White |
| Dimensions (L x W x H) | 10 x 10 x 18 cm (4 x 4 x 7 in.) |
| Disposable | Single-use |
| Height (Metric) | 18 cm |
| Length (Metric) | 10 cm |
| Size | Small |
| Sterility | Non-sterile |
| Width (Metric) | 10 cm |
| Show More |