missing translation for 'onlineSavingsMsg'
Learn More

Brady™ White Tyvek™ Tags

Catalog No. 19112957
Change view
Click to view available options
Dimensions (L x W):
1.375 x 2.75 in.
2.125 x 4.25 in.
2.375 x 4.75 in.
2.625 x 5.25 in.
2.875 x 5.75 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19112957 1.375 x 2.75 in.
19112958 2.125 x 4.25 in.
19112960 2.625 x 5.25 in.
19112959 2.375 x 4.75 in.
19112961 2.875 x 5.75 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19112957 Supplier Brady™ Supplier No. 102032

Identify facility equipment to your exact specifications. Brady™ White Tyvek™ Tags are durable one-sided tags with a write-on area that accepts most marking pens.

  • Tyvek™ tags are a lightweight, tear proof and weather resistant alternative to cardstock tags
  • They provide more durability but still offer the flexibility to write your own custom message on the spot
  • Metal grommet prevents tearing from the point of attachment
  • Tag includes a write-on area that accepts most marking pens
  • Tags are appropriate for indoor or outdoor use

Specifications

Quantity 1000/Pk.
Color White
Material Tyvek
Shape Rectangle
Dimensions (L x W) 1.375 x 2.75 in.
Label Type Not laminated