missing translation for 'onlineSavingsMsg'
Learn More
Learn More
International Enviroguard™ White SMS Coveralls with Elastic Wrists and Open Ankles
SMS coveralls provide three layers of protection against particulates such as dirt and dust, and light liquid splashes or sprays. Lightweight and breathable fabric allows heat to escape so wearers stay cool.
Supplier: International Enviroguard™ 22223XL
Description
- SMS coveralls are lightweight and breathable, so wearers stay cool throughout long shifts
- Durable, three-layer SMS fabric (Spunbond Meltblown Spunbond) protects against light liquid splashes and sprays and provides an ideal particulate barrier
- Storm flap covers the zipper to help keep non-hazardous particles and liquids from entering zipper seams
- Wider, gusseted leg area gives wearer more room to move in all directions without ripping fabric
- Elastic wrists offer a secure fit to help keep sleeves in place to prevent skin exposure
- Elastic back stretches with user for more comfort and better tear resistance
- Possible applications include asbestos and lead abatement, mold remediation, manufacturing, assembly operations
Specifications
| General Purpose Coveralls | |
| Unisex | |
| White | |
| Elastic at Back | |
| Elastic | |
| Standard Collar | |
| Serged | |
| Liquid, Particle | |
| 2222 | |
| Asbestos and Lead Abatement, Mold Remediation, Manufacturing, Assembly Operations |
| 3X-Large | |
| SMS Fabric | |
| Single-use | |
| Open | |
| Collared | |
| Front Zipper with Storm Flap | |
| Non-sterile | |
| Yes | |
| None |