Learn More
International Enviroguard™ White SMS Coveralls with Elastic Wrists and Ankles
SMS coveralls provide three layers of protection against particulates such as dirt and dust, and light liquid splashes or sprays. Lightweight and breathable fabric allows heat to escape so wearers stay cool.
Supplier:  International Enviroguard™ 22235XL
 View more versions of this product
Catalog No. 17-184-210
Only null left
Explore available promotions
Description
Description
  • SMS coveralls are lightweight and breathable, so wearers stay cool throughout long shifts
  • Durable, three-layer SMS fabric (Spunbond Meltblown Spunbond) protects against light liquid splashes and sprays and provides an ideal particulate barrier
  • Storm flap covers the zipper to help keep non-hazardous particles and liquids from entering zipper seams
  • Wider, gusseted leg area gives wearer more room to move in all directions without ripping fabric
  • Elastic wrists and ankles offer a secure fit to prevent skin exposure
  • Elastic back stretches with user for more comfort and better tear resistance
  • Possible applications include asbestos and lead abatement, mold remediation, manufacturing, assembly operations
Specifications
Specifications
General Purpose Coveralls
Unisex
White
Elastic at Back
Elastic
Standard Collar
Serged
Liquid, Particle
2223
Asbestos and Lead Abatement, Mold Remediation, Manufacturing, Assembly Operations
5X-Large
SMS Fabric
Single-use
Elastic
Collared
Front Zipper with Storm Flap
Non-sterile
Yes
None
Product Suggestions
Product Suggestions