missing translation for 'onlineSavingsMsg'
Learn More

Spilfyter™ White Scrim Wiper

Catalog No. 17000273
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17000273 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17000273 Supplier Spilfyter™ Supplier No. C3350W
Only null left

The Spilfyter White Scrim Wiper remains strong even when wet and offers excellent glass and surface cleaning.

  • Low linting, excellent for glass and surface cleaning
  • 150 interfolded pop out wipers in a dispenser box, 10 dispenser boxes per case
  • Cellulose-based wiper with a nylon 'Scrim' netting in the middle to provide wet strength and absorbency
  • Soft feel, won't scratch most surfaces
  • Use for small liquid clean-up and detail wiping

Specifications

Material Scrim (Cellulose Based)
Color White
Container Type Corrugate Box
Disposable Single-use
Dimensions (L x W) 9.25 x 12.5 in. (23.5 x 31.75 cm)
For Use With (Application) Cleaning
Packaging Quantity 150 Wipes/Dispenser, 10 Boxes/Cs.
Sterility Non-sterile