Learn More
Fisherbrand™ White Polypropylene Caps
Catalog No. FB02912064A
Change view
Click to view available options
Neck Finish:
13-425
48-400
70-400
Product Type:
Lined Cap
Unlined Cap
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish Product Type
FB02912064A 13-425 Lined Cap
FB02912291A 48-400 Lined Cap
FB02912332A 48-400 Unlined Cap
FB02912088A 70-400 Lined Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. FB02912064A
Fisherbrand™ White Polypropylene Caps include a clear writing surface for easy identification.
  • Foamed polyethylene liner provides tight seal and chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products
  • PTFE faced foamed polyethylene liner offers chemical resistance excellent for environmental samples, pharmaceuticals and diagnostic reagents
  • Poly-vinyl liner provides an excellent moisture barrier good for mild acids, alkalis, solvents, alcohols, oils and aqueous products
Specifications
Color White
Material Polypropylene
Quantity 500
Liner Material PTFE/Polyethylene Foam
Neck Finish 13-425
Product Type Lined Cap