Learn More
Fisherbrand™ White Polypropylene Caps
White polypropylene screw caps provides clear surface for writing on cap
Description
- Foamed polyethylene liner provides tight seal and chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products
- PTFE faced foamed polyethylene liner offers chemical resistance excellent for environmental samples, pharmaceuticals and diagnostic reagents
- Poly-vinyl liner provides an excellent moisture barrier good for mild acids, alkalis, solvents, alcohols, oils and aqueous products
Chemical Identifiers
| White | |
| Polypropylene | |
| Lined Cap |
Specifications
| White | |
| 144/Cs. | |
| 70-400 |
| Polypropylene | |
| PTFE/Polyethylene Foam | |
| Lined Cap |