missing translation for 'onlineSavingsMsg'
Learn More
Learn More
- Home
- Products
- Stoppers and Closures
- Bottle Caps
- Fisherbrand™ White Polypropylene Caps
Fisherbrand™ White Polypropylene Caps
Catalog No. FB02912075
Change view
Click to view available options
Neck Finish:
15-425
18-400
20-400
22-400
24-400
28-400
33-400
38-400
43-400
45-400
48-400
53-400
63-400
70-400
83-400
89-400
Liner Material:
Polyethylene Foam
PTFE/Polyethylene Foam
Pulp/Vinyl
Quantity:
100/Cs.
1300/Cs.
144/Cs.
2100/Cs.
288/Cs.
370/Cs.
500/Cs.
22 product options available for selection
| Catalog No. | Neck Finish | Liner Material | Quantity |
|---|---|---|---|
| FB02912075 | 48-400 | PTFE/Polyethylene Foam | 100/Cs. |
| FB02911836A | 33-400 | PTFE/Polyethylene Foam | 2100/Cs. |
| FB02911844A | 53-400 | Pulp/Vinyl | 1300/Cs. |
| FB02912066 | 18-400 | PTFE/Polyethylene Foam | 100/Cs. |
| FB02912065 | 15-425 | PTFE/Polyethylene Foam | 500/Cs. |
| FB02912067 | 20-400 | PTFE/Polyethylene Foam | 100/Cs. |
| FB02912069 | 24-400 | PTFE/Polyethylene Foam | 100/Cs. |
| FB02912068 | 22-400 | PTFE/Polyethylene Foam | 500/Cs. |
| FB02912070 | 28-400 | PTFE/Polyethylene Foam | 100/Cs. |
| FB02912071 | 33-400 | PTFE/Polyethylene Foam | 100/Cs. |
| FB02912072 | 38-400 | PTFE/Polyethylene Foam | 500/Cs. |
| FB02912073 | 43-400 | PTFE/Polyethylene Foam | 500/Cs. |
| FB02912074 | 45-400 | PTFE/Polyethylene Foam | 500/Cs. |
| FB02912076 | 53-400 | PTFE/Polyethylene Foam | 100/Cs. |
| FB02912078 | 70-400 | PTFE/Polyethylene Foam | 144/Cs. |
| FB02912079 | 83-400 | PTFE/Polyethylene Foam | 144/Cs. |
| FB02912080 | 89-400 | PTFE/Polyethylene Foam | 144/Cs. |
| FB02912085 | 63-400 | PTFE/Polyethylene Foam | 288/Cs. |
| FB02912093A | 53-400 | Polyethylene Foam | 1300/Cs. |
| FB02912099 | 53-400 | Polyethylene Foam | 500/Cs. |
| FB02912094A | 89-400 | Polyethylene Foam | 370/Cs. |
| FB02912292A | 53-400 | Pulp/Vinyl | 288/Cs. |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
Description
- Foamed polyethylene liner provides tight seal and chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products
- PTFE faced foamed polyethylene liner offers chemical resistance excellent for environmental samples, pharmaceuticals and diagnostic reagents
- Poly-vinyl liner provides an excellent moisture barrier good for mild acids, alkalis, solvents, alcohols, oils and aqueous products
Specifications
Specifications
| Color | White |
| Material | Polypropylene |
| Quantity | 100/Cs. |
| Liner Material | PTFE/Polyethylene Foam |
| Neck Finish | 48-400 |
| Product Type | Lined Cap |