missing translation for 'onlineSavingsMsg'
Learn More

Qorpak™ White Metal Pulp/Polyethylene Lined Caps

Exhibit tolerance to widest range of temperatures

$739.94 - $5507.97

Specifications

Color White
Material Metal
Liner Material Pulp/Polyethylene
For Use With (Equipment) Wide mouth bottle
View More Specs

Products 2
Catalog Number Mfr. No. Neck Finish No. per Case Packaging Price Quantity  
Catalog Number Mfr. No. Neck Finish No. per Case Packaging Price Quantity  
02-993-380
View Documents
Qorpak™
CAP06662
70-400 144 Cs. Bagged
Case of 144 Each for $739.94
Only null left
 
02-991-338
View Documents
Qorpak™
CAP00435
28-400 5400 Cs. Bulk
Case of 5400 Each for $5,507.97
Only null left
 
Description

Description

  • Resistant to fracture from impact
  • Offered in 400 GPI thread finish
  • For general-purpose
  • Ideal for powders, non-acidic food products and consumer products

Specifications

Specifications

White
Metal
Pulp/Polyethylene
Wide mouth bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'