missing translation for 'onlineSavingsMsg'
Learn More

Bullard™ White Cleanable Carriage Body Straps

Catalog No. 19087975
Change view
Click to view available options
For use with: SALUS HC System
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19087975 For use with: SALUS HC System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19087975 Supplier Bullard™ Supplier No. SALHCBS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

White cleanable replacement carriage body straps.

  • White
  • Replacement carriage body straps

Specifications

Color White
Disposable Reusable
Product Type Carriage Straps
For Use With (Equipment) SALUS HC System
Model SALHCBS