missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ White/Blue Performance Crew Socks

Catalog No. 1139520
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
1139520 Large
1139516 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1139520 Supplier Wells Lamont™ Supplier No. 8501LN

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

White or blue socks, constructed of polyester/nylon/spandex, are ideal for cleanrooms.

  • Men's Performance Work Socks provide support and protection for your feet while you work.
  • The mesh upper foot is designed for ventilation and moisture evaporation, keeping your feet dry throughout the day.
  • Socks feature reinforced heel and toe as well as an Achilles heel pad for added protection and comfort.
  • The socks non-binding top do not slip down and provide all-day gentle comfort.
  • Added compression at the arch provides added support and stability.

Specifications

Size Large
For Use With (Application) Cleanroom Use
Material Polyester, Nylon, Spandex
Color White, Blue