missing translation for 'onlineSavingsMsg'
Learn More
Learn More
International Enviroguard™ White Biohazard Coveralls with Attached Boots
ViroGuard 2 disposable biohazard suits are designed with fabric and seams that have been rigorously tested for protection against blood and bloodborne pathogens.
Supplier: International Enviroguard™ W25013XL
Description
- Durable fabric is built to withstand tears and abrasions in tough working environments
- Both fabric and seams pass ASTM F1670 and F1671 tests for protection against blood, bodily fluids, and bloodborne pathogens
- Zipper front with a sealable storm flap helps keep contaminants from entering or seeping through otherwise-exposed zipper seams
- Reinforced with double-sided tape at the wrists, your gloves will remain secure as you work
- Red fabric pull tab on the zipper makes it quick and easy to find so you can suit up or down in no time
- Taped seams seal off seams from contaminants, while elastic wrists and ankles keep coveralls secure and in place to avoid skin exposure
- For added convenience, thumb and finger loops are included to help prevent sleeves from riding up
- Attached boots protect your feet while latex-free, elastic wrists offer a secure fit to prevent skin exposure of the arms
- Mandarin collar with hook and loop closure at neck provides a better neck seal
- Roomy, one-piece raglan sleeves resist tears around shoulders
- Gusseted crotch gives the user more freedom to move, bend, crouch, climb, and squat without ripping fabric
- Possible applications include emergency response, healthcare, category 3 black water, sanitation work, crime scene remediation, and animal research
Specifications
| A50 Breathable Particle and Splash Protection Coveralls | |
| Unisex | |
| White | |
| Elastic at Back | |
| Elastic, Finger Loops | |
| Standard | |
| Serged, Taped | |
| Bloodborne Pathogens, Liquid, Particle, Tears | |
| None |
| 3X-Large | |
| Multi-layer Barrier Laminate Fabric | |
| Single-use | |
| Attached Boots | |
| Collared | |
| 2-way Zipper with Sealed Storm Flap | |
| Non-sterile | |
| Yes |