missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ White 96-Well Immuno Plates
GREENER_CHOICE

Catalog No. 14245196Q
Change view
Click to view available options
Well Shape:
Round, Flat Bottom (F-well)
Surface Treatment:
MaxiSorp
Microfluor 1
Microfluor 2
Microlite 1+
Microlite 2+
PolySorp
Universal Binding
Volume (Metric) Working:
280 μL
330 μL
350 μL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Shape Surface Treatment Volume (Metric) Working
14245196Q Round, Flat Bottom (F-well) Microlite 2+ 280 μL
1256605 Round, Flat Bottom (F-well) MaxiSorp 350 μL
1256606 Round, Flat Bottom (F-well) PolySorp 350 μL
14245196P Round, Flat Bottom (F-well) Microlite 1+ 280 μL
14245197B Round, Flat Bottom (F-well) Microfluor 1 280 μL
14245178 Round, Flat Bottom (F-well) Microfluor 2 280 μL
21377273 Round, Flat Bottom (F-well) Universal Binding 330 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 14245196Q Supplier Thermo Scientific™ Supplier No. 7572
Only null left

Conduct quantitative and qualitative immunoassays like ELISA and binding assays with these white, 96-well immuno plates.

Conduct quantitative and qualitative immunoassays, like ELISA and binding assays, with Thermo Scientific™ White 96-Well Immuno Plates. These plates, which are compatible with common instruments, deliver optimal results, lot-to-lot reliability and well-to-well reproducibility.

Features

  • Plate has SBS footprint — fits standard equipment
  • Available with flat- or round-bottom wells
  • Choice of three surface types for optimal binding
  • Certified binding homogeneity/reproducibility

Choice of Surface:

  • Thermo Scientific PolySorp, Microfluor 1, Microlite 1+ and Universal Binding (UB) for adsorption of hydrophobic molecules
  • Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays
  • Thermo Scientific Microfluor 2 and Microlite 2+ are slightly hydrophilic and bind a diverse range of biomolecules

White:

  • Gives maximum reflection
  • Minimum autofluorescence and autoluminescence

Compatible with:

Lids for Thermo Scientific MicroWell plates, sealing tape and breathable membrane

Ordering Information:

Availability may vary by country.

Specifications

Binding Property Slightly Hydrophilic
Format Standard
Lid With Lid
Detection Method Chemiluminescence
Array 8 x 12
Packaging 50 Cases
Color White
Product Type Microplate
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 280 μL
Surface Treatment Microlite 2+
Well Shape Round, Flat Bottom (F-well)
For Use With (Application) Luminescence
No. per Case 50
No. per Pack 10
Plate Blocking None
Target Molecule Antibodies (Glycoproteins) and Proteins
Show More Show Less