missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ White 96-Well Immuno Plates

Description
Conduct quantitative and qualitative immunoassays, like ELISA and binding assays, with Thermo Scientific™ White 96-Well Immuno Plates. These plates, which are compatible with common instruments, deliver optimal results, lot-to-lot reliability and well-to-well reproducibility.
Features
- Plate has SBS footprint — fits standard equipment
- Available with flat- or round-bottom wells
- Choice of three surface types for optimal binding
- Certified binding homogeneity/reproducibility
Choice of Surface:
- Thermo Scientific PolySorp, Microfluor 1, Microlite 1+ and Universal Binding (UB) for adsorption of hydrophobic molecules
- Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays
- Thermo Scientific Microfluor 2 and Microlite 2+ are slightly hydrophilic and bind a diverse range of biomolecules
White:
- Gives maximum reflection
- Minimum autofluorescence and autoluminescence
Compatible with:
Lids for Thermo Scientific MicroWell plates, sealing tape and breathable membrane
Ordering Information:
Availability may vary by country.
Specifications
Specifications
| Binding Property | Hydrophobic |
| Format | Standard |
| Lid | With Lid |
| Detection Method | Chemiluminescence |
| Array | 8 x 12 |
| Packaging | 50 Cases |
| Binding Type | Adsorption |
| Color | White |
| Product Type | Microplate |
| Material | Polystyrene |
| Show More |