Learn More
Thermo Scientific™ White 96-Well Immuno Plates
Conduct quantitative and qualitative immunoassays like ELISA and binding assays with these white, 96-well immuno plates.
Supplier:  Thermo Scientific™ 436111
 Afficher plus de versions de ce produit
Numéro de catalogue. 1256606
Il en reste null
Description
Description
Conduct quantitative and qualitative immunoassays, like ELISA and binding assays, with Thermo Scientific™ White 96-Well Immuno Plates. These plates, which are compatible with common instruments, deliver optimal results, lot-to-lot reliability and well-to-well reproducibility.
Features
  • Plate has SBS footprint — fits standard equipment
  • Available with flat- or round-bottom wells
  • Choice of three surface types for optimal binding
  • Certified binding homogeneity/reproducibility
Choice of Surface:
  • Thermo Scientific PolySorp, Microfluor 1, Microlite 1+ and Universal Binding (UB) for adsorption of hydrophobic molecules
  • Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays
  • Thermo Scientific Microfluor 2 and Microlite 2+ are slightly hydrophilic and bind a diverse range of biomolecules
White:
  • Gives maximum reflection
  • Minimum autofluorescence and autoluminescence
Compatible with:
Lids for Thermo Scientific MicroWell plates, sealing tape and breathable membrane
Ordering Information:
Availability may vary by country.
Spécifications
Identifiants chimiques
Standard
White
96
8 x 12
Microplate
Spécifications
Hydrophobic
With Lid
8 x 12
White
Polystyrene
350 μL
PolySorp
Fluorescence and Luminescence
10
80/Cs.
Standard
Chemiluminescence
80 Cases
Microplate
96
Non-sterile
Round, Flat Bottom (F-well)
80
None
Lipids
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS